missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli CFA / I fimbrial subunit E Protein
Click to view available options
Quantity:
1 mg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
1mg
200µg
500µg
50µg
Specifications
Specifications
| Accession Number | WP_001033658.1 |
| Name | CFA / I fimbrial subunit E Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not determined. |
| Gene Symbol | cfaE |
| Protein Length | Full Length |
| Product Type | Recombinant Protein |
| Cross Reactivity | E. coli |
| Species | E. coli |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction