missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Bifunctional protein HldE Protein
A DNA sequence encoding the Escherichia coli (strain K12) Bifunctional protein HldE, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6949-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417524.1 | |
Bifunctional protein HldE Protein | |
Research Use Only | |
hldE | |
Recombinant Protein | |
E. coli | |
MKVTLPEFER AGVMVVGDVM LDRYWYGPTS RISPEAPVPV VKVNTIEERP GGAANVAMNI ASLGANARLV GLTGIDDAAR ALSKSLADVN VKCDFVSVPT HPTITKLRVL SRNQQLIRLD FEEGFEGVDP QPLHERINQA LSSIGALVLS DYAKGALASV QQMIQLARKA GVPVLIDPKG TDFERYRGAT LLTPNLSEFE AVVGKCKTEE EIVERGMKLI ADYELSALLV TRSEQGMSLL QPGKAPLHMP TQAQEVYDVT GAGDTVIGVL AATLAAGNSL EEACFFANAA AGVVVGKLGT STVSPIELEN AVRGRADTGF GVMTEEELKL AVAAARKRGE KVVMTNGVFD ILHAGHVSYL ANARKLGDRL IVAVNSDAST KRLKGDSRPV NPLEQRMIVL GALEAVDWVV SFEEDTPQRL IAGILPDLLV KGGDYKPEEI AGSKEVWANG GEVLVLNFED GCSTTNIIKK IQQDKKG | |
Greater than 90% as determined by SDS-PAGE. |
947548 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |