Learn More
enQuireBio™ Recombinant E. coli Bifunctional protein HldE Protein
A DNA sequence encoding the Escherichia coli (strain K12) Bifunctional protein HldE, was expressed in the hosts and tags indicated.
440.00€ - 3745.00€
Specifications
Accession Number | NP_417524.1 |
---|---|
Gene ID (Entrez) | 947548 |
Name | Bifunctional protein HldE Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096830
|
enQuireBio™
QP6949-10UG |
10 μg |
440.00€
10µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16036840
|
enQuireBio™
QP6949-50UG |
50 μg |
630.00€
50µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16086830
|
enQuireBio™
QP6949-100UG |
100 μg |
1070.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16016840
|
enQuireBio™
QP6949-200UG |
200 μg |
1765.00€
200µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16026840
|
enQuireBio™
QP6949-500UG |
500 μg |
2360.00€
500µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16006840
|
enQuireBio™
QP6949-1MG |
1 mg |
3745.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Specifications
NP_417524.1 | |
Bifunctional protein HldE Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
947548 | |
Research Use Only | |
hldE | |
Recombinant Protein | |
E. coli | |
MKVTLPEFER AGVMVVGDVM LDRYWYGPTS RISPEAPVPV VKVNTIEERP GGAANVAMNI ASLGANARLV GLTGIDDAAR ALSKSLADVN VKCDFVSVPT HPTITKLRVL SRNQQLIRLD FEEGFEGVDP QPLHERINQA LSSIGALVLS DYAKGALASV QQMIQLARKA GVPVLIDPKG TDFERYRGAT LLTPNLSEFE AVVGKCKTEE EIVERGMKLI ADYELSALLV TRSEQGMSLL QPGKAPLHMP TQAQEVYDVT GAGDTVIGVL AATLAAGNSL EEACFFANAA AGVVVGKLGT STVSPIELEN AVRGRADTGF GVMTEEELKL AVAAARKRGE KVVMTNGVFD ILHAGHVSYL ANARKLGDRL IVAVNSDAST KRLKGDSRPV NPLEQRMIVL GALEAVDWVV SFEEDTPQRL IAGILPDLLV KGGDYKPEEI AGSKEVWANG GEVLVLNFED GCSTTNIIKK IQQDKKG | |
Greater than 90% as determined by SDS-PAGE. |