missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase RecG Protein
A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase RecG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7112-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418109.1 | |
ATP-dependent DNA helicase RecG Protein | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
948162 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
recG | |
Recombinant Protein | |
E. coli | |
MKGRLLDAVP LSSLTGVGAA LSNKLAKINL HTVQDLLLHL PLRYEDRTHL YPIGELLPGV YATVEGEVLN CNISFGGRRM MTCQISDGSG ILTMRFFNFS AAMKNSLAAG RRVLAYGEAK RGKYGAEMIH PEYRVQGDLS TPELQETLTP VYPTTEGVKQ ATLRKLTDQA LDLLDTCAIE ELLPPELSQG MMTLPEALRT LHRPPPTLQL SDLETGQHPA QRRLILEELL AHNLSMLALR AGAQRFHAQP LSANDTLKNK LLAALPFKPT GAQARVVAEI ERDMALDVPM MRLVQGDVGS GKTLVAALAA LRAIAHGKQV ALMAPTELLA EQHANNFRNW FAPLGIEVGW LAGKQKGKAR LAQQEAIASG QVQMIVGTHA IFQEQVQFNG LALVIIDEQH RFGVHQRLAL WEKGQQQGFH PHQLIMTATP IPRTLAMTAY ADLDTSVIDE LPPGRTPVTT VAIPDTRRTD IIDRVHHACI TEGRQAYWVC TLIEESELLE AQAAEATWEE LKLALPELNV GLVHGRMKPA EKQAVMASFK QGELHLLVAT TVIEVGVDVP NASLMIIENP ERLGLAQLHQ LRGRVGRGAV ASHCVLLYKT PLSKTAQIRL QVLRDSNDGF VIAQKDLEIR GPGELLGTRQ TGNAEFKVAD LLRDQAMIPE VQRLARHIHE RYPQQAKALI ERWMPETERY SNA |