missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog IL-8 / CXCL8 Protein
A DNA sequence encoding the Canis familiaris (Dog) (Canis lupus familiaris) IL-8 / CXCL8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6227-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003200.1 | |
IL-8 / CXCL8 Protein | |
Research Use Only | |
CXCL8 | |
Recombinant Protein | |
Yeast | |
AVLSRVSSEL RCQCIKTHST PFHPKYIKEL RVIDSGPHCE NSEIIVKLFN GNEVCLDPKE KWVQKVVQIF LKKAEKQDP | |
Greater than 90% as determined by SDS-PAGE. |
403850 | |
200 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |