missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog IL-8 / CXCL8 Protein
A DNA sequence encoding the Canis familiaris (Dog) (Canis lupus familiaris) IL-8 / CXCL8, was expressed in the hosts and tags indicated.
352.00€ - 4075.00€
Specifications
Accession Number | NP_001003200.1 |
---|---|
Gene ID (Entrez) | 403850 |
Name | IL-8 / CXCL8 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011760
|
enQuireBio™
QP6227-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16031760
|
enQuireBio™
QP6227-YE-1MG |
1 mg |
4075.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16021760
|
enQuireBio™
QP6227-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041760
|
enQuireBio™
QP6227-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16051760
|
enQuireBio™
QP6227-YE-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061760
|
enQuireBio™
QP6227-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001003200.1 | |
IL-8 / CXCL8 Protein | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |
403850 | |
Research Use Only | |
CXCL8 | |
Recombinant Protein | |
Yeast | |
AVLSRVSSEL RCQCIKTHST PFHPKYIKEL RVIDSGPHCE NSEIIVKLFN GNEVCLDPKE KWVQKVVQIF LKKAEKQDP | |
Greater than 90% as determined by SDS-PAGE. |