missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cytomegalovirus 65 kDa phosphoProtein
A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) 65 kDa phosphoprotein, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Name | 65 kDa phosphoProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | UL83 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008470
|
enQuireBio™
QP7365-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16028470
|
enQuireBio™
QP7365-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16018470
|
enQuireBio™
QP7365-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038470
|
enQuireBio™
QP7365-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048470
|
enQuireBio™
QP7365-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058470
|
enQuireBio™
QP7365-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
65 kDa phosphoProtein | |
Not determined. | |
Partial (see sequence information for more details). | |
Cytomegalovirus | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
UL83 | |
Recombinant Protein | |
E. coli | |
FDIDLLLQRG PQYSEHPTFT SQYRIQGKLE YRHTWDRHDE GAAQGDDDVW TSGSDSDEEL VTTERKTPRV TGGGAMAGAS TSAGRKRKSA SSATACTSGV MTRGRLKAES TVAPEEDTDE DSDNEIHNPA | |
Greater than 90% as determined by SDS-PAGE. |