missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus 65 kDa phosphoProtein

A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) 65 kDa phosphoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7365-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038470

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

65 kDa phosphoProtein
Research Use Only
UL83
Recombinant Protein
E. coli
FDIDLLLQRG PQYSEHPTFT SQYRIQGKLE YRHTWDRHDE GAAQGDDDVW TSGSDSDEEL VTTERKTPRV TGGGAMAGAS TSAGRKRKSA SSATACTSGV MTRGRLKAES TVAPEEDTDE DSDNEIHNPA
Greater than 90% as determined by SDS-PAGE.
200 μg
Not determined.
Partial (see sequence information for more details).
Cytomegalovirus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers