missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Monkey Nectin-2/CD112 His Protein, CF
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus DNAM-1/CD226 Fc Chimera (Catalog # 9276-DN ) is immobilized at 1 μg/mL, 100 uL/well, Recombinant Cynomolgus Monkey Nectin-2/CD112 His-tag Protein binds with an ED50 of 0.4-2.4 μg/mL.
Specifications
Specifications
| Accession Number | XP_005589607.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 5819 |
| Molecular Weight (g/mol) | M.W. (Observed): 41-50 kDa, under reducing conditions, M.W. (Theoretical): 36 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Nectin-2/CD112 protein Gln32-Gly360, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HVEB, HVEBpoliovirus receptor-like 2, MPH, nectin 2, Nectin2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2, PRR2nectin-2, PVRL2, PVRR2poliovirus receptor related 2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction