missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Interferon tau-1 Protein
A DNA sequence encoding the Bos taurus (Bovine) Interferon tau-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7076-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001015511.3 | |
Interferon tau-1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
317698 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
IFNT1 | |
Recombinant Protein | |
E. coli | |
CYLSEDHMLG ARENLRLLAR MNRLSPHPCL QDRKDFGLPQ EMVEGNQLQK DQAISVLHEM LQQCFNLFYT EHSSAAWNTT LLEQLCTGLQ QQLEDLDACL GPVMGEKDSD MGRMGPILTV KKYFQGIHVY LKEKEYSDCA WEIIRVEMMR ALSSSTTLQK RLRKMGGDLN SL |