missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Hemagglutinin-esterase Protein
A DNA sequence encoding the Bos taurus (Bovine) Hemagglutinin-esterase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9549-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
Hemagglutinin-esterase Protein | |
Research Use Only | |
HE | |
Recombinant Protein | |
Yeast | |
FDNPPTNVVS HLNGDWFLFG DSRSDCNHVV NTNPRNYSYM DLNPALCDSG KISSKAGNSI FRSFHFTDFY NYTGEGQQII FYEGVNFTPY HAFKCTTSGS NDIWMQNKGL FYTQVYKNMA VYRSLTFVNV PYVYNGSAQS TALCKSGSLV LNNPAYIARE ANFGDYYYKV EADFYLSGCD EYIVPLCIFN GKFLSNTKYY DDSQYYFNKD TGVIYGLNST ETITTGFDFN CHYLVLPSGN YLAISNELLL TVPTKAICLN KRKDFTPVQV VDSRWNNARQ SDNMTAVACQ PPYCYFRNST TNYVGVYDIN HGDAGFTSIL SGLLYDSPCF SQQGVFRYDN VSSVWPLYSY GRCPTAADIN TPDVPICVYD PLPLILLGIL LGVAVIIIVV LLLYFMVDNG TRLHDA | |
Greater than 81.30% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |