missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Hemagglutinin-esterase Protein
A DNA sequence encoding the Bos taurus (Bovine) Hemagglutinin-esterase, was expressed in the hosts and tags indicated.
352.00€ - 3175.00€
Specifications
Name | Hemagglutinin-esterase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | HE |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038461
|
enQuireBio™
QP9549-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16078461
|
enQuireBio™
QP9549-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16048461
|
enQuireBio™
QP9549-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16068461
|
enQuireBio™
QP9549-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16088461
|
enQuireBio™
QP9549-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058461
|
enQuireBio™
QP9549-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Hemagglutinin-esterase Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
HE | |
Recombinant Protein | |
Yeast | |
FDNPPTNVVS HLNGDWFLFG DSRSDCNHVV NTNPRNYSYM DLNPALCDSG KISSKAGNSI FRSFHFTDFY NYTGEGQQII FYEGVNFTPY HAFKCTTSGS NDIWMQNKGL FYTQVYKNMA VYRSLTFVNV PYVYNGSAQS TALCKSGSLV LNNPAYIARE ANFGDYYYKV EADFYLSGCD EYIVPLCIFN GKFLSNTKYY DDSQYYFNKD TGVIYGLNST ETITTGFDFN CHYLVLPSGN YLAISNELLL TVPTKAICLN KRKDFTPVQV VDSRWNNARQ SDNMTAVACQ PPYCYFRNST TNYVGVYDIN HGDAGFTSIL SGLLYDSPCF SQQGVFRYDN VSSVWPLYSY GRCPTAADIN TPDVPICVYD PLPLILLGIL LGVAVIIIVV LLLYFMVDNG TRLHDA | |
Greater than 81.30% as determined by SDS-PAGE. |