missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Bovine Enteropeptidase/Enterokinase Protein

Product Code. 18668678 Shop All Bio Techne Products
Click to view available options
Quantity:
100 IU
250 IU
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18668678

Brand: Novus Biologicals™ NBP261359100IU

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enzyme Activity
Formulation 50 mM Tris-HCl, pH 8.0, 0.5 M NaCl
Gene ID (Entrez) 5651
Molecular Weight (g/mol) MolecularWeight-theroretical: 28 kDa
Quantity 100 IU
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of Enteropeptidase/Enterokinase as determined by LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15
Gene Symbol TMPRSS15
Research Category Epitope Tags, Immunology, Innate Immunity
Purity or Quality Grade Peptide affinity purified
Protein Enteropeptidase/Enterokinase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.