missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Recombinant B18R Protein
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
Description
- The B18R protein, which is encoded by the B18R open reading frame in the Western Reserve (WR) strain of vaccinia virus, is a type I interferon (IFN)-binding protein that is known to inhibit IFN antiviral response (Colamonici et al.) B18R protein has been shown to exhibit neutralizing activity to type I interferon family members (Meyers et al.) and to protect cells from the antiviral effects of interferon (Vancová et al.) It was recently shown that HIV-1–dependent induction of type I IFN activity and BAFF protein levels was abrogated by the soluble B18R protein, with no effect observed on the cell viability (Gomez et al.) B18R protein has been reported to aid in cellular reprogramming using RNA-mediated gene delivery (Warren et al.; Yoshioka N et al.)
Specifications
Specifications
| Name | Type I IFN inhibitor, Vaccinia virus-encoded neutralizing type I interferon receptor |
| Quantity | 50 μg |
| Cell Type | Pluripotent Stem Cells |
| Purity or Quality Grade | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction