missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant A. tubingensis PNGase Protein

Product Code. 18710789
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18710789 20 μg 20µg
18768609 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18710789 Supplier R&D Systems™ Supplier No. 9586GH020
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

rFm PNGase F is also available!

Measured by its ability to deglycosylate ribonuclease B under denatured conditions. 50% ribonuclease B (6 μg) is deglycosylated by 500 ng of Recombinant A.tubingensis PNGase within 120 minutes, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number OJI88926.1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) M.W. (Observed): 72-85 kDa, reducing conditions; M.W. (theoretical): 60 kDa
Quantity 20 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived a. tubingensis PNGase At protein Leu22-Ser557 (F78I, E352G, P381S, S406Q, T412S, S414F, S451T and S555T), with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species A. tubingensis
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PNGase At
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.