Learn More
IKA™ RCT Basic
Product Code. 17779657 Shop All IKA Products
Change view
Click to view available options
Speed:
50 to 1500 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Unit Size
17779657 50 to 1500 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 17779657 Supplier IKA™ Supplier No. 0025005927
  • ONLINE EXCLUSIVE PRICE
    1103.34€
    1025.00€ / Pack of 1
    Save 78.34€
    7% Off
In Stock
Add to basket
Edge
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
RCT basic is suitable for stirring tasks up to 20 L (H2O) and reaches a hotplate temperature of up to 310™C. With the connection option for an external temperature sensor (PT 1000.60 included in delivery), the temperature can be measured and controlled directly in the reaction medium.
  • Tempered shatterproof glass surface for optimal chemical resistance and easy cleaning
  • Largest display in its class with easy-to-read LED display
  • Illuminated symbols for displaying important status information (set and actual temperature, heating status, temperature sensor
  • USB and RS232 interface for control or documentation of the test parameters via a PC
  • Compatible with labworldsoft™ laboratory software
  • QR code for quick access to device information, accessories, downloads and extended warranty
  • Easily accessible main switch on the front of the device
  • Safe, robust and compliant with standards
  • RCT basic contains the proven safety features for IKA magnetic stirrers. In accordance with the DIN EN 61010-2-010 standard, it meets all safety requirements for laboratory equipment for heating substances and is therefore also suitable for unattended operation.
  • Tested and certified by TÜV SÜD according to standard IEC 61010-1 (cTÜVus)
  • Adjustable hotplate temperature safety circuit (with tool)
  • Confirmation mode (operating mode D) prevents the unintentional change of the set setpoints. At restart, the confirmation of the safety temperature is necessary.
  • Confirmation mode (operating mode D) prevents the unintentional change of the set setpoints. At restart, the confirmation of the safety temperature is necessary.
  • Automatic switch-off of the temperature control function if the connected external temperature sensor is not immersed in the medium or is defective. Function selectable, timeout time adjustable (Error 5).
  • Enclosed design (protection class IP42) guarantees long service life, even under extreme conditions in the laboratory
  • Reliable operation even with cold media. Extended temperature display down to -20°C (with external sensor).
  • Protected electronic connections on the back of the device
  • Fireproof die-cast aluminum housing with high quality and durable powder coating
  • DIN socket 12878 for connecting an electronic contact thermometer, e.g. ETS-D5 for high-presence temperature control. In this combination, the experimental setup is extended by a further independent safety circuit for the reaction medium.
  • Proven technology
  • Heating plate made of aluminum for optimal and homogeneous heat transfer
  • Excellent magnetic coupling
  • Soft start prevents magnetic stir bars from breaking off during the start phase
  • Two optimized temperature control modes ensure fast heating or precise temperature control without overshooting
  • Push and turn buttons for independent adjustment of the setpoints and starting / stopping of temperature and speed
TRUSTED_SUSTAINABILITY
Specifications
Speed 50 to 1500 rpm
Stirring Positions 1
Top Plate Material Aluminum Alloy
Display LED
Voltage 220/230 V
Height (Metric) 85 mm
Top Plate Diameter (Metric) 135 mm
Width (Metric) 160 mm
Frequency 50/60 Hz
Certifications/Compliance Documents for qualification LAB, Factory Calibration Certificate LAB, DQ/IQ/OQ/PQ Documentation LAB, General Certificates LAB (2.2 oder 3.1), Qualification Execution (IQ/OQ/PQ) LAB
Warranty and Service Offering 2 plus 1 Year Warranty
Includes IKAFLON™ 30 Magnetic Stirring Bar, IKAFLON™ 40 Magnetic Stirring Bar, Screw Driver, Stainless Steel PT 1000.60 Temperature Sensor
Temperature Range (Metric) 0°C to 310°C
Type Magnetic Stirrer
Weight (Metric) 2.4 kg