missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCOR1/CoREST Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58094-25ul
This item is not returnable.
View return policy
Description
RCOR1/CoREST Polyclonal specifically detects RCOR1/CoREST in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| RCOR1/CoREST | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| KIAA0071CORESTRCORREST corepressor, Protein CoREST, REST corepressor 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| RCOR1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QEWEAEHGKEETNGPSNQKPVKSPDNSIKMPEEEDEAPVLDVRYASAS | |
| 25 μL | |
| Transcription Factors and Regulators | |
| 23186 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido