missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPMS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15187-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the RRM family of RNA-binding proteins. The RRM domain is between 80-100 amino acids in length and family members contain one to four copies of the domain. The RRM domain consists of two short stretches of conserved sequence called RNP1 and RNP2, as well as a few highly conserved hydrophobic residues. The protein encoded by this gene has a single, putative RRM domain in its N-terminus. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| RBPMS | |
| Polyclonal | |
| Unconjugated | |
| RBPMS | |
| HERMES, RBP MS, RBPMS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11030, 19663, 498642 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q93062, Q9WVB0 | |
| Rbpms | |
| RBPMS Fusion Protein Ag7256 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction