missing translation for 'onlineSavingsMsg'
Learn More

RBP4/Retinol-Binding Protein 4 Antibody (423619), Allophycocyanin/Cy7, R&D Systems™

Product Code. 30035510 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30035510 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30035510 Supplier Novus Biologicals Supplier No. FAB34761APCCY7
  • ONLINE EXCLUSIVE PRICE
    674.00€
    637.35€ / 0.1mL
    Save 36.65€
    5% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications Flow Cytometry
Classification Monoclonal
Clone 423619
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4, Met1-Leu201, Accession # NP_035385
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5950
Target Species Human, Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.