Learn More
RBP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16852954 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16852954 20 μL 20µL
16842954 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16852954 Supplier Proteintech Supplier No. 143521AP20UL
Rabbit Polyclonal Antibody
IRBP shuttles 11-cis and all trans retinoids between the retinol isomerase in the pigment epithelium and the visual pigments in the photoreceptor cells of the retina.
Specifications
Antigen RBP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBP3
Gene Accession No. P10745, P49194
Gene Alias D10S64, D10S65, D10S66, IRBP, RBP3, RBPI, Retinol binding protein 3
Gene Symbols RBP3
Host Species Rabbit
Immunogen RBP3 Fusion Protein Ag5699
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19661, 24711, 5949
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG