missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM4B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93750-0.02ml
This item is not returnable.
View return policy
Description
RBM4B Polyclonal antibody specifically detects RBM4B in Human samples. It is validated for Western Blot
Specifications
| RBM4B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC10871, RBM30, RBM4L, RNA binding motif protein 30, RNA binding motif protein 4B, RNA-binding motif protein 30, RNA-binding motif protein 4B, RNA-binding protein 30, RNA-binding protein 4B, ZCCHC15, ZCRB3B, zinc finger CCHC-type and RNA binding motif 3B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-359 of human RBM4B (NP_113680.1). RMHVQLSTSRLRTAPGMGDQSGCYRCGKEGHWSKECPVDRTGRVADFTEQYNEQYGAVRTPYTMGYGESMYYNDAYGALDYYKRYRVRSYEAVAAAAAASAYNYAEQTMSHLPQVQSTTVTSHLNSTSVDPYDRHLLPNSGAAATSAAMAAAAATTSSYYGRDRSPLRRAAAMLPTVGEGYGYGPESELSQASAATRNSLYDMARYEREQYVDRARYSAF | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 83759 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction