missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM48 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80470
This item is not returnable.
View return policy
Description
RBM48 Polyclonal specifically detects RBM48 in Human samples. It is validated for Western Blot.
Specifications
| C7orf64 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C7orf64, chromosome 7 open reading frame 64, DKFZp564O0523, DKFZp686D1651, HSPC304, hypothetical protein LOC84060, RNA binding motif protein 48 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84060 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_115496 | |
| RBM48 | |
| Synthetic peptide directed towards the C terminal of human DKFZP564O0523. Peptide sequence FLQTNPTGNEIMIGPLLPDISKVDMHDDSLNTTANLIRHKLKEVISSVPK. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Sumatran orangutan: 100%; Human: 100%; Rat: 92%; Mouse: 85%; Chicken: 78%. | |
| Human, Mouse, Rat, Pig, Canine, Equine, Guinea Pig, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction