missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP4/RbAp48 Antibody (C.15200033), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62798
This item is not returnable.
View return policy
Description
RBBP4/RbAp48 Monoclonal specifically detects RBBP4/RbAp48 in Human samples. It is validated for Western Blot.
Specifications
| RBBP4/RbAp48 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide and 0.05% ProClin 300 | |
| RBBP4 | |
| Human RbAp48 protein (retinoblastoma binding protein p48) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| C.15200033 | |
| Western Blot 1:1000 | |
| CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48 | |
| Mouse | |
| Protein G purified | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors | |
| 5928 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction