Learn More
RB1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30374-1-AP-150UL
This item is not returnable.
View return policy
Description
The RB1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human RB1. This antibody recognizes human antigen. The RB1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RB1 | |
| 1300 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| OSRC, p105 Rb, pp110, pRb, RB, RB1, retinoblastoma 1 | |
| 150 μL | |
| 5925 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |