Learn More
Rat μ Opioid R/OPRM1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6866-SP
This item is not returnable.
View return policy
Description
mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Western Blot.
Specifications
| mu Opioid R/OPRM1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P33535 | |
| OPRM1 | |
| Synthetic peptide corresponding to His385-Pro398 of rat μ Opioid R/OPRM1 Accession # P33535 | |
| 25 μg | |
| Primary | |
| Detects rat μ Opioid R/OPRM1 in direct ELISAs. | |
| Rat | |
| Purified |
| Western Blot | |
| 677014 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4988 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |