missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG2B PE-conjugated Antibody, R&D Systems™
Description
IgG2b Monoclonal specifically detects IgG2b in Rat samples. It is validated for Control.
Specifications
Specifications
| Antigen | IgG2b |
| Classification | Monoclonal |
| Clone | 141945 |
| Conjugate | R-PE |
| Dilution | Control 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Host Species | Rat |
| Immunogen | KLH |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Quantity | 200 Tests |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?