missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG Horseradish Peroxidase-conjugated Antibody, R&D Systems™

Product Code. 18171191
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18171191 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18171191 Supplier R&D Systems Supplier No. HAF005
  • ONLINE EXCLUSIVE PRICE
    272.83€
    242.55€ / 1mL
    Save 30.28€
    11% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

IgG Polyclonal specifically detects IgG in Rat samples. It is validated for ELISA∣Immunohistochemistry∣Simple Western∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications ELISA, Immunohistochemistry, Simple Western, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Goat
Immunogen Rat IgG
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects rat IgG 1 IgG 2A IgG 2B heavy and light chains in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with human IgG and rabbit IgG is observed.
Target Species Rat
Content And Storage Do not freeze. 6 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.