missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tollip, Rat, Clone: Kimmy-1, Abnova™
Rat monoclonal antibody raised against recombinant Tollip.
Brand: Abnova MAB3709.100ug
This item is not returnable.
View return policy
Specifications
| Tollip | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Tollip | |
| Tollip | |
| Recombinant protein corresponding to mouse Tollip. | |
| RUO | |
| 54473 | |
| Human, Mouse | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Kimmy-1 | |
| Rat monoclonal antibody raised against recombinant Tollip. | |
| In PBS (0.02% sodium azide) | |
| 4930403G24Rik/4931428G15Rik | |
| Rat | |
| 100 μg | |
| Primary | |
| Recognizes human and mouse Tollip. | |
| Store at 4°C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction