missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG3, Clone: R40-82, FITC, Secondary Antibody, BD

Product Code. 15817208
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15817208 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817208 Supplier BD Biosciences Supplier No. 553403
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The R40-82 antibody reacts specifically with mouse IgG3 of all strains. It does not react with other Ig isotypes.

Host Species: Rat
Clone: R40-82
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Mouse Pooled Ig

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgG3
Classification Monoclonal
Clone R40-82
Concentration 0.5mg/mL
Conjugate FITC
Host Species Rat
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.