missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgD, Clone: 11-26c.2a, Secondary Antibody, BD

Product Code. 15887218
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15887218 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15887218 Supplier BD Biosciences Supplier No. 553438
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The 11-26c.2a antibody reacts with mouse immunoglobulin D of all Igh-C haplotypes (e.g., IgDa, IgDb, IgDe), and it does not react with other immunoglobulin isotypes. Although 11-26c.2a mAb binds membrane IgD expressed on the splenic B-cell surface with hi

Host Species: Rat
Clone: 11-26c.2a
Isotype: IgG2a, κ
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse IgD
Classification Monoclonal
Clone 11-26C.2A
Concentration 0.5mg/mL
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.