missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse Ig κ Light Chain, Clone: 187.1, Secondary Antibody, BD

Product Code. 15806168
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15806168 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15806168 Supplier BD Biosciences Supplier No. 550003
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The 187.1 antibody reacts specifically with mouse Igs bearing κ light chain. It does not react with λ1 or λ2 light chain or heavy chain. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 187.1
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse IgG2b κ secreted by MPC-11 plasmacytoma

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat anti-Mouse Ig κ Light Chain
Classification Monoclonal
Clone 187.1
Concentration 0.5mg/mL
Conjugate FITC
Host Species Rat
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.