missing translation for 'onlineSavingsMsg'
Learn More

CD83 Rat anti-Mouse, BUV395, Clone: Michel-19, BD Optibuild™

Rat Monoclonal Antibody

Brand:  BD Biosciences 740323

Product Code. 15826689

  • ONLINE EXCLUSIVE PRICE
    442.00€
    441.74€ / Each
    Save 0.26€
    0.06% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Michel-19 antibody reacts with CD83, a member of the immunoglobulin superfamily that is expressed on mature dendritic cells and activated T lymphocytes. Furthermore, thymic cortical epithelial cells express Cd83 transcripts. CD83 is involved in the regulation of T-cell development and immune responses, and its ligand is found on a subpopulation of splenic B lymphocytes.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD83
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse CD83 Recombinant Protein
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
Michel-19
Brilliant Ultraviolet 395
Rat
Affinity Purified
RUO
Mouse
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only