missing translation for 'onlineSavingsMsg'
Learn More

Lyl1, Rat, Clone: KT43, Abnova™

Product Code. 16130120
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16130120 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16130120 Supplier Abnova Supplier No. MAB1675.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat monoclonal antibody raised against synthetic peptide of Lyl1.

Specifications

Antigen Lyl1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KT43
Conjugate Unconjugated
Description Rat monoclonal antibody raised against synthetic peptide of Lyl1.
Dilution Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Lyl1
Gene Alias Lyl-1/bHLHa18
Gene Symbols Lyl1
Host Species Rat
Immunogen A synthetic peptide (conjugated with BSA) corresponding to mouse Lyl1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17095
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.