missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP, Rat, Clone: 1A5, Abnova™
Rat monoclonal antibody raised against recombinant GFP.
Brand: Abnova MAB6746.100ug
This item is not returnable.
View return policy
Specifications
| GFP | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rat | |
| 100 μg | |
| Primary | |
| Jellyfish | |
| Liquid |
| ChIP Assay, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 1A5 | |
| Rat monoclonal antibody raised against recombinant GFP. | |
| In PBS (50% glycerol) | |
| Recombinant protein corresponding to Aequorea victoria GFP. | |
| RUO | |
| This antibody is specific to GFP and GFP-fused proteins. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction