Learn More
AID Rat anti-Human, Alexa Fluor 647, Clone: EK2-5G9, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 565785
Description
The EK2-5G9 monoclonal antibody specifically binds to human Activation-induced cytidine deaminase (AICDA), also known as AID, or Single-stranded DNA cytosine deaminase. AID is an enzyme that preferentially acts upon certain DNA sequence motifs, such as those located in immunoglobulin variable regions, to induce cytidine-to-uracil mutations. AID-induced mutations initiate both somatic hypermutation (SHM) and class switch recombination (CSR) of the immunoglobulin genes in activated B cells during immune responses. While essential for immunoglobulin gene diversification, dysregulated AID activity can result in genomic instability and oncogenic transformation. Originally thought to be B-cell specific, AID now appears to be abnormally expressed in several epithelial cancers. AID may also play roles in DNA demethylation.
Specifications
| AID | |
| Monoclonal | |
| 0.2mg/mL | |
| AID; ARP2; CDA2; HEL-S-284; HIGM2; Cytidine aminohydrolase | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
| EK2-5G9 | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human AID (185-198) Peptide | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.