missing translation for 'onlineSavingsMsg'
Learn More

RASA4 Antibody, Novus Biologicals™

Product Code. 18138811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18138811 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18138811 Supplier Novus Biologicals Supplier No. NBP215090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RASA4 Polyclonal specifically detects RASA4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASA4
Applications Western Blot
Classification Polyclonal
Concentration 0.71mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O43374
Gene Alias Ca2+-promoted Ras inactivator, Calcium-promoted Ras inactivator, CAPRIGAPLKIAA0538RASA4B, FLJ59070, Putative Ras GTPase-activating protein 4B, ras GTPase-activating protein 4, RAS p21 protein activator 4MGC131890, RasGAP-activating-like protein 2
Gene Symbols RASA4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human RASA4. The exact sequence is proprietary.
Molecular Weight of Antigen 90 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10156
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.