missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Rap1 Activation Assay Kit, Human, Mouse, Rat, 20 Test
Description
- Application: Measure RAP1B
- Sample types: Tissue, Suspension cells, Adherent cells
- Species reactivity: Mouse, Rat, Human
- Citations: 5
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Western Blot Kit |
| Detection Location | In-Solution Detection |
| Format | Kit |
| No. of Reactions | 20 Tests |
| Reactivity | Human, Mouse, Rat |
| Target Molecule | RAP1B (Human, Mouse, Rat) |
| Detection Method | Colorimetric |
| Cross Reactivity | Human, Mouse, Rat |
| Sample Type | Tissue, Suspension Cells, Adherent Cells |
| Quantity | 20 Tests |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?