Learn More
R&D Systems™ ELISA Plate-coating Buffer
Product Code. 30021823
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30021823 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30021823 Supplier R&D Systems™ Supplier No. DY006
ELISA Plate-coating Buffer from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The ELISA Plate-coating Buffer quantifies Buffers, Chelators and Reagents.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) ELISA
Quantity 250 mL