Learn More
Thermo Scientific™ MarqMetrix Replacement FlowCell BallProbe
Product Code. 17759417 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Replacement FlowCell BallProbe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17759417 Replacement FlowCell BallProbe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17759417 Supplier Thermo Scientific™ Supplier No. 8100286101
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
TRUSTED_SUSTAINABILITY
Spezifikation
For Use With (Equipment) MarqMetrix All-In-One Process Raman Analyzer
Type Replacement FlowCell BallProbe