missing translation for 'onlineSavingsMsg'
Learn More

RALY Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68011-1-IG-20UL

 View more versions of this product

Product Code. 30234924

  • ONLINE EXCLUSIVE PRICE
    145.00€
    129.00€ / 20µL
    Save 16.00€
    11% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RALY antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RALY. This antibody recognizes human antigen. The RALY antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.
Specifications

Specifications

RALY
Monoclonal
1000 μg/mL
HNRPCL2; MGC117312; P542
Recombinant Protein
20 μL
Primary
Human
30-37 kDa
Liquid
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA
2B10C3
Unconjugated
Mouse
Protein G purification
RUO
22913
Store at -20°C. Stable for one year after shipment.
Monoclonal Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.