missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAIDD/CRADD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77048-0.025mg
This item is not returnable.
View return policy
Description
RAIDD/CRADD Polyclonal specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RAIDD/CRADD | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain | |
| Rabbit | |
| 18 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer, Cell Biology, Tumor Suppressors | |
| 8738 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| AAB42217 | |
| CRADD | |
| Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human RAIDD. The immunogen is located within the last 50 amino acids of RAIDD. Amino Acid Squence: HNGLRAVEVDPSLLLHMLE | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction