Learn More
Invitrogen™ RAE1 alpha/beta/delta/epsilon Monoclonal Antibody (199205)
GREENER_CHOICE
Product Code. 30533643
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30533643 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30533643 Supplier Invitrogen™ Supplier No. MA561040
Rat Monoclonal Antibody
Reconstitute at 0.5 mg/mL in sterile PBS.
RAE1 (RNA Export 1) is a gene that plays a pivotal role in the nuclear export of mRNA in eukaryotic cells. This gene encodes a protein that is integral to the mRNA export machinery, ensuring the proper transport of mRNA from the nucleus to the cytoplasm for translation. The gene has several isoforms, including RAE1 alpha, beta, delta, and epsilon, which may have unique or overlapping functions in different tissues or developmental stages. The proper functioning of RAE1 and its isoforms is crucial for maintaining normal cellular processes and gene expression. Disruptions or mutations in the RAE1 gene can lead to significant defects in mRNA export, potentially causing a range of diseases, including neurodevelopmental disorders and cancers. Ongoing research into RAE1 and its various isoforms is essential for a deeper understanding of mRNA transport mechanisms and their broader implications in cellular health and disease.
TRUSTED_SUSTAINABILITY
Specifications
Antigen RAE1 alpha/beta/delta/epsilon
Applications Flow Cytometry, Inhibition Assays
Classification Monoclonal
Clone 199205
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with Rae-1 beta (Accession # NP_033043) and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 (Accession # Q9JI58).
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2a