missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51D Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93383-0.1ml
This item is not returnable.
View return policy
Description
Rad51D Polyclonal antibody specifically detects Rad51D in Human samples. It is validated for Western Blot
Specifications
| Rad51D | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| DNA repair protein RAD51 homolog 4, HsTRAD, R51H3Trad, RAD51 homolog D, RAD51DRAD51 (S. cerevisiae)-like 3, RAD51-like 3 (S. cerevisiae), RAD51-like protein 3, recombination repair protein, TRAD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-328 of human Rad51D (NP_002869.3). MGVLRVGLCPGLTEEMIQLLRSHRIKTVVDLVSADLEEVAQKCGLSYKALVALRRVLLAQFSAFPVNGADLYEELKTSTAILSTGIGSLDKLLDAGLYTGEVTEIVGGPGSGKTQVCLCMAANVAHGLQQNVLYVDSNGGLTASRLLQLLQAKTQDEEEQAEALRRIQVVHAFDIFQMLDVLQELRGTVAQQVTGSSGTVKVVVVDSVTAVVSPLLGGQQREGLALMMQLARELKTLARDLGMAVVVTNHITRDRDSGRLKPALGRSWSFVPSTRILLDTIEGAGASGGRRMACLAKSSRQPTGFQEMVDIGTWGTSEQSATLQGDQT | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
| 5892 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction