missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 593.00€
Specifications
| Antigen | Rad51C |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18226502
|
Novus Biologicals
NBP2-55278 |
100 μL |
593.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611969
|
Novus Biologicals
NBP2-55278-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rad51C Polyclonal specifically detects Rad51C in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Rad51C | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 5889 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MRGKTFRFEMQRDLVSFPLSPAVRVKLVSAGFQTAEELLEVKPSELSKEVGISKAEALETLQIIRRECLTNKPRYAGTSESHKKCTALELLEQEHTQGFIITFCSALDDILGGGVPLMKTTEICGAPGVG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
| RAD51C | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title