missing translation for 'onlineSavingsMsg'
Learn More

rac-Sertraline Hydrochloride ((1RS,4RS)-4-(3,4-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride), Mikromol™

Product Code. 30052407
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052407 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052407 Supplier Mikromol Supplier No. MM0317.160025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

rac-Sertraline Hydrochloride ((1RS,4RS)-4-(3,4-Dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-cis-Sertraline Hydrochloride
CAS 79617-89-3
Analyte or Component Names rac-cis-Sertraline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H17 Cl2 N . Cl H
Certifications/Compliance ISO 17025
Synonym (1R,4R)-rel-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride (1:1), cis-N-Methyl-4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthaleneamine hydrochloride, cis-(+/-)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride, (+/-)-Sertraline hydrochloride
InChI Formula InChI=1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1
SMILES Cl.CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13
IUPAC Name (1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
Molecular Weight (g/mol) 342.69
Quantity 25 mg
Formula Weight 341.0505 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.