Learn More
rac-Norsertraline Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Norsertraline hydrochloride |
| CAS | 91797-57-8 |
| Analyte or Component Names | Norsertraline hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 Cl2 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (±)-cis-N-Desmethylsertraline Hydrochloride, cis-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, (±)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrochloride, rac-Norsertraline Hydrochloride |
| InChI Formula | InChI=1S/C16H15Cl2N.ClH/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13;/h1-5,7,9,11,16H,6,8,19H2;1H/t11-,16-;/m0./s1 |
| Solution Type | Neat |
| SMILES | Cl.N[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13 |
| Show More |