Learn More
rac Metoprolol, TRC
Product Code. 17939984
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939984 100 mg 100mg
30137660 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17939984 Supplier Toronto Research Chemicals Supplier No. TRCM338815100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51384-51-1
Formula Weight 267.1834
InChI Formula InChI=1S/C15H25NO3/c1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3/h4-7,12,14,16-17H,8-11H2,1-3H3/t14-/m1/s1
IUPAC Name (2R)-1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molecular Formula C15 H25 N O3
Molecular Weight (g/mol) 267.36
Percent Purity >95%
SMILES COCCc1ccc(OC[C@H](O)CNC(C)C)cc1
Synonym (RS)-Metoprolol, (±)-Metoprolol, 1-Isopropylamino-3-[4-(2-methoxyethyl)phenoxy]-2-propanol, 1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol, Beatrolol, CGP 2175, Meijoprolol, Metocard, Metohexal, Metoprolol, Seroken, Spesicor, Toprol, dl-Metoprolol, 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-
Chemical Name or Material (R)-Metoprolol
Grade HPLC