missing translation for 'onlineSavingsMsg'
Learn More

rac-1-t-Boc-3-methyl-4[N-(1-oxopropyl)-N-(phenylamino]isonipecotic Acid Methyl Ester, TRC

Product Code. 30346112
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346112 1 mg 1mg
30336407 10 mg 10mg
30346559 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30346112 Supplier Toronto Research Chemicals Supplier No. TRCB6569901MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-1-t-Boc-3-methyl-4[N-(1-oxopropyl)-N-phenylamino]isonipecotic Acid Methyl Ester
Analyte or Component Names rac-1-t-Boc-3-methyl-4[N-(1-oxopropyl)-N-phenylamino]isonipecotic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C22 H32 N2 O5
InChI Formula InChI=1S/C22H32N2O5/c1-7-18(25)24(17-11-9-8-10-12-17)22(19(26)28-6)13-14-23(15-16(22)2)20(27)29-21(3,4)5/h8-12,16H,7,13-15H2,1-6H3/t16-,22+/m0/s1
Solution Type Neat
SMILES CCC(=O)N(c1ccccc1)[C@@]2(CCN(C[C@@H]2C)C(=O)OC(C)(C)C)C(=O)OC
IUPAC Name 1-O-tert-butyl 4-O-methyl (3S,4R)-3-methyl-4-(N-propanoylanilino)piperidine-1,4-dicarboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 404.5
Quantity 1 mg
Formula Weight 404.231
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.