missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17816-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the RAB gene family which belongs to the RAS GTPase superfamily. The proteins in the family of RAS-related signaling molecules are small GTP-binding proteins that play important roles in the regulation of exocytotic and endocytotic pathways. This gene maps to the site of an ancestral telomere fusion event and may be a subtelomeric gene. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| RABL2A | |
| Polyclonal | |
| Unconjugated | |
| RABL2A | |
| Rab like protein 2A, RABL2A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11159, 362987, 68708 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E9Q9D5, Q9UBK7 | |
| Rabl2, Rabl2A | |
| RABL2A Fusion Protein Ag12135 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction