missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Rabies Virus Glycoprotein Antibody (rafivirumab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28279-50ug
This item is not returnable.
View return policy
Description
Rabies Virus Glycoprotein Monoclonal antibody specifically detects Rabies Virus Glycoprotein in Virus samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Rabies Virus Glycoprotein | |
| Monoclonal | |
| Unconjugated | |
| Human | |
| Protein A purified | |
| RUO | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| rafivirumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Rabies virus GP | |
| 50 μg | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction